Learn More
Sino Biological Recombinant Mouse CD146/MCAM Protein (His Tag)
Supplier:  Sino Biological 50794M08H50
Encompass_Preferred
A DNA sequence encoding the mouse MCAM (Q8R2Y2-1) extracellular domain (Met 1-Val 563) was expressed, fused with a C-terminal polyhistidine tag.
Catalog No. 50-224-7302