Learn More
Sino Biological Mouse Cd1d1 & B2M Heterodimer Protein (His Tag)
Catalog No. 502993093
Encompass_Preferred
A DNA sequence encoding Mouse Cd1d1 (NP_031665.2) (Met1-Gly305) was fused with a polyhistidine tag at the C-terminus, constructed the plasmid 1; A DNA sequence encoding the Mouse B2M (P01887) (Met1-Met119, A105D) constructed the plasmid 2. The two plasmids were co-expressed and the Mouse Cd1d1/B2M heterodimer was purified.
Catalog No. 50-299-3093 Supplier Sino Biological Supplier No. CT070M08H150
A DNA sequence encoding Mouse Cd1d1 (NP_031665.2) (Met1-Gly305) was fused with a polyhistidine tag at the C-terminus, constructed the plasmid 1; A DNA sequence encoding the Mouse B2M (P01887) (Met1-Met119, A105D) constructed the plasmid 2. The two plasmids were co-expressed and the Mouse Cd1d1/B2M heterodimer was purified.