Learn More
Sino Biological Recombinant Anti-CD200 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50074R002100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD200 (rM CD200; Catalog#50074-M08H; O54901.1; Met 1-Gly 232).
Catalog No. 50-255-3485