Learn More
Sino Biological Recombinant Anti-CD200 Antibody (APC), Rabbit Monoclonal
Supplier:  Sino Biological 50074R002A25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD200 (rM CD200; Catalog#50074-M08H; O54901.1; Met1-Gly232) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-255-3483