Learn More
Sino Biological Mouse CD200 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 50074RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CD200 (rM CD200; Catalog#50074-M08H; O54901.1; Met 1-Gly 232). CD200 specific IgG was purified by mouse CD200 affinity chromatography.
Catalog No. 50-237-2838