Learn More
Sino Biological Recombinant Anti-CD200R Antibody (APC), Rabbit Monoclonal
Supplier:  Sino Biological 50209R181A100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD200R1 (rM CD200R1; Catalog#50209-M08H; NP_067300.1; Met1-Pro238) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-255-3377