Learn More
Sino Biological Rhesus Siglec-2 / CD22 Protein (His Tag) 1mg
Supplier:  Sino Biological 90246C08H1
Encompass_Preferred
A DNA sequence encoding the rhesus CD22 (EHH29920.1) (Met1-Arg686) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-222-2697