Learn More
Sino Biological Recombinant Anti-CD27/TNFRSF7 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50110R034100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD27 (rM CD27; Catalog#50110-M08H; NP_001028298.1; Met 1-Arg 182).
Catalog No. 50-240-8095