Learn More
Sino Biological Anti-CD27/TNFRSF7 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50110RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CD27 (rM CD27; Catalog#50110-M08H; NP_001028298.1; Met 1-Arg 182). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-1198