Learn More
Sino Biological Anti-CD27/TNFRSF7 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50110T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CD27 (rM CD27; Catalog#50110-M08H; NP_001028298.1; Met1-Arg182). CD27 specific IgG was purified by Mouse CD27 affinity chromatography.
Catalog No. 50-255-3456