Learn More
Sino Biological Recombinant Anti-PD-L1/B7-H1 Antibody (PE), Rabbit Monoclonal
Supplier:  Sino Biological 50010R485P25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse PD-L1 / B7-H1 / CD274 (rM PD-L1 / B7-H1 / CD274; Catalog#50010-M08H; NP_068693.1; Met1-Thr238) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-240-8502