Learn More
Sino Biological Anti-PD-L1/B7-H1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50010RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse B7-H1 / PD-L1 extracellular domain (rM B7-H1; Catalog#50010-M08H; NP_068693.1; Met 1-Thr 238). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8863