Learn More
Sino Biological Anti-PD-L1/B7-H1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50010T32100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse PD-L1/B7-H1/CD274 (rM PD-L1/B7-H1/CD274; Catalog#50010-M08H; NP_068693.1; Met1-Thr238). PD-L1/B7-H1/CD274 specific IgG was purified by Mouse PD-L1/B7-H1/CD274 affinity chromatography.
Catalog No. 50-241-1662