Learn More
Sino Biological Recombinant Mouse PD-L1/B7-H1 Protein (ECD, His & hFc Tag)
Supplier:  Sino Biological 50010M03H1
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Thr 238) of mouse PD-L1 (NP_068693.1) was fused with the C-terminal His-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-255-3558