Learn More
Sino Biological Recombinant Mouse PD-L1/B7-H1 Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 50010M08H1
Encompass_Preferred
A DNA sequence encoding the mouse CD274 (NP_068693.1) extracellular domain (Met 1-Thr 238) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-3556