Learn More
Sino Biological Recombinant Mouse PD-L1/B7-H1 Protein (His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 50010M49HB20
Encompass_Preferred
A DNA sequence encoding the Mouse CD274 (NP_068693.1)(Met1-Thr238) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-241-1374