Learn More
Sino Biological Recombinant Mouse B7-H3 Protein (His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 50973M49HB100
Encompass_Preferred
A DNA sequence encoding the Mouse CD276 (NP_598744.1)(Met1-Phe244) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-255-3041