Learn More
Sino Biological Anti-CD28 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50103RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CD28 (rM CD28; Catalog#50103-M08H; NP_031668.3; Met 1-Lys 149). CD28 specific IgG was purified by Mouse CD28 affinity chromatography.
Catalog No. 50-240-8327