Learn More
Sino Biological Mouse CD28 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 50103RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CD28 / TP44 (Catalog#50103-M08H; NP_031668.3; Met1-Lys149). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-237-3569