Learn More
Sino Biological Recombinant Mouse CD28 Protein (ECD, Dimer, hFc Tag), HPLC-verified
Supplier:  Sino Biological 50103M02H100
Encompass_Preferred
A DNA sequence encoding the mouse CD28 (NP_031668.3) (Met1-Lys149) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-255-3461