Learn More
Sino Biological Recombinant Mouse CD28 Protein (His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 50103M08HB100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Lys 149) of mouse CD28 (NP_031668.3) was fused with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-255-3460