Learn More
ACROBiosystems Monoclonal Anti-Human CD3 Antibody, Mouse IgG2a, Clone OKT3, Premium Grade, 1 mg
Catalog No. 502253790
Encompass_Preferred
The Monoclonal Anti-Human CD3 Antibody, Mouse IgG2a (Clone: OKT3) is a premium grade, recombinantly produced antibody derived from HEK293 cells. It specifically targets the CD3 epsilon molecule, a key component of the T cell receptor-CD3 complex involved in antigen recognition and intracellular signaling. Designed for early-stage discovery and preclinical applications, this product offers a seamless transition to GMP-grade versions, featuring an identical cell clone, production process, and quality control profile.
  • Supplied as a 0.2 μm filtered solution in PBS, pH 7.4 with trehalose
  • Purity >95% as determined by SDS-PAGE and SEC-HPLC
  • Endotoxin levels <0.002 EU per μg
  • Rigorous quality control including sterility and mycoplasma testing
  • Demonstrated bioactivity with a typical EC50 of 0.12 ng/mL for IL-2 stimulation
This item is not returnable. View return policy
Catalog No. 50-225-3790 Supplier ACROBiosystems Supplier No. CDEM120A1MG
The Monoclonal Anti-Human CD3 Antibody, Mouse IgG2a (Clone: OKT3) is a premium grade, recombinantly produced antibody derived from HEK293 cells. It specifically targets the CD3 epsilon molecule, a key component of the T cell receptor-CD3 complex involved in antigen recognition and intracellular signaling. Designed for early-stage discovery and preclinical applications, this product offers a seamless transition to GMP-grade versions, featuring an identical cell clone, production process, and quality control profile.
  • Supplied as a 0.2 μm filtered solution in PBS, pH 7.4 with trehalose
  • Purity >95% as determined by SDS-PAGE and SEC-HPLC
  • Endotoxin levels <0.002 EU per μg
  • Rigorous quality control including sterility and mycoplasma testing
  • Demonstrated bioactivity with a typical EC50 of 0.12 ng/mL for IL-2 stimulation