Learn More
ACROBiosystems Mouse CD3 50ug
Catalog No. 502535317
Encompass_Preferred
Biotinylated Anti-Human CD3 Antibody Mouse IgG2aAvitag (Clone: OKT3) premium grade is a chimeric monoclonal antibody recombinantly expressed from human 293 cells (HEK293) which combines the variable region of a mouse monoclonal antibody. The mouse monoclonal antibody is produced from a hybridoma resulting from fusion of SP2/0 myeloma and B-lymphocytes obtained from a mouse immunized with CD3 epsilon.
Catalog No. 50-253-5317 Supplier ACROBiosystems Supplier No. CDEBV84G050UG
Biotinylated Anti-Human CD3 Antibody Mouse IgG2aAvitag (Clone: OKT3) premium grade is a chimeric monoclonal antibody recombinantly expressed from human 293 cells (HEK293) which combines the variable region of a mouse monoclonal antibody. The mouse monoclonal antibody is produced from a hybridoma resulting from fusion of SP2/0 myeloma and B-lymphocytes obtained from a mouse immunized with CD3 epsilon.