Learn More
ACROBiosystems ACROBiosystems Mouse CD3 epsilon&CD3 gamma Heterodimer Protein,Fc,His Tag&Fc,Flag Tag (MALS verified)
Supplier:  ACROBiosystems CDGM52W21MG
Encompass_Preferred
Recombinant Protein;1MG;Mouse CD3E&CD3G Heterodimer Protein,Fc,His Tag&Fc,Flag Tag (CDG-M52W2) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 108 (CD3E) & Gln 23 - Ser 116 (CD3G) (Accession # P22646-1 (CD3E) & P11942-1 (CD3G)).;Mouse CD3E&CD3G Heterodimer Protein,Fc,His Tag&Fc,Flag Tag is produced by co-expression of CD3E and CD3G, has a calculated MW of 41.5 kDa (CD3E) and 41.6 kDa (CD3G). Subunit CD3E is fused with a human IgG1 Fc tag at the C-terminus, followed by a polyhistidine tag and subunit CD3G is fused with a human IgG1 Fc tag at the C-terminus, followed by a Flag tag. As a result of glycosylation, the protein migrates as 48-55 kDa under reducing (R) condition, and 100 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Mouse CD3E&CD3G Heterodimer Protein,Fc,His Tag&Fc,Flag Tag (Cat. No. CDG-M52W2) at 1 μg/mL (100 μL/well) can bind Biotinylated CD3e Monoclonal Antibody (Clone : 145-2C11) with a linear range of 0.2-6 ng/mL (QC tested).;Immobi
Catalog No. 50-210-7410