Learn More
ACROBiosystems Recombinant Protein;Mouse CD3 epsilon Protein, Fc Tag;HEK293;100UG;CDE,FLJ18683,T3E,TCRE,CD3E,CD3-epsilon,CD3 epsilon ,CDE-H822R
Supplier: ACROBiosystems CDEM5256100UG
Recombinant Protein;Mouse CD3 epsilon Protein, Fc Tag;100UG;Mouse CD3E, Fc Tag (CDE-M5256) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 108 (Accession # P22646).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 36 kDa. The protein migrates as 45 kDa under reducing (R) condition (SDS-PAGE).