Learn More
Medchemexpress LLC Mouse CD3+ T Cells Negative Selection Kit | 1 ML
SDP
Catalog No. 5000431537
Encompass_Preferred
The Mouse CD3+ T Cells Negative Selection Kit is designed for the isolation of CD3+ T cells from single cell suspensions of mouse spleen cells and lymph nodes. It employs a biotin-labeled monoclonal antibody to mark non-target cells, which are subsequently removed using streptavidin-labeled magnetic beads. This user-friendly, portable kit provides high-purity cells suitable for various downstream applications.
  • Enables sorting of target cells in as little as 15 minutes.
  • Eliminates the need for separation columns by using a magnetic separator for swift cell separation.
  • Can yield sorted cells with a purity exceeding 95%.
  • Sorted cells are free of antibody and magnetic bead labeling, and retain high activity and functionality for downstream experiments.
Catalog No. 50-004-31537 Supplier Medchemexpress LLC Supplier No. HYK03061ML
Add to Cart
Add to Cart
The Mouse CD3+ T Cells Negative Selection Kit is designed for the isolation of CD3+ T cells from single cell suspensions of mouse spleen cells and lymph nodes. It employs a biotin-labeled monoclonal antibody to mark non-target cells, which are subsequently removed using streptavidin-labeled magnetic beads. This user-friendly, portable kit provides high-purity cells suitable for various downstream applications.
  • Enables sorting of target cells in as little as 15 minutes.
  • Eliminates the need for separation columns by using a magnetic separator for swift cell separation.
  • Can yield sorted cells with a purity exceeding 95%.
  • Sorted cells are free of antibody and magnetic bead labeling, and retain high activity and functionality for downstream experiments.