Learn More
Sino Biological Anti-CD300E Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 107559T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Mouse CD300E, and purified by antigen affinity chromatography.
Catalog No. 50-241-1690