Learn More
Sino Biological Anti-Siglec-3/CD33 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50712RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CD33 / Siglec-3 (Catalog#50712-M08H; Q63994-1; Met1-Glu240). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-1563