Learn More
Sino Biological Anti-Siglec-3/CD33 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50712T48100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse Siglec-3/CD33 (rM Siglec-3/CD33; Catalog#50712-M08H; Q63994-1; Met1-Glu240). Siglec-3/CD33 specific IgG was purified by Mouse Siglec-3/CD33 affinity chromatography.
Catalog No. 50-255-3128