Learn More
Sino Biological Recombinant Anti-CD34 Antibody (PerCP), Rabbit Monoclonal
Supplier:  Sino Biological 50589R013C100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD34 (rM CD34; Catalog#50589-M08H; NP_001104529.1; Met 1-Thr 287) and conjugated with PerCP under optimum conditions, the unreacted PerCP was removed.
Catalog No. 50-240-8242