Learn More
Sino Biological Recombinant Anti-CD34 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50589R305100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD34 (Catalog#50589-M08H; NP_001104529.1; Met1-Thr287).
Catalog No. 50-241-1836