Learn More
Sino Biological Mouse CD34 Protein 500ug (Sino Biological)
Supplier:  Sino Biological 50709R348A25
Encompass_Preferred
A DNA sequence encoding the Mouse CD34 (NP_001104529.1) (Met1-Thr287) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-4513