Learn More
Sino Biological Anti-CD36 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50422T26100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CD36/SCARB3 (rM CD36/SCARB3; Catalog#50422-M08H; NP_001153030.1; Gly30-Lys439). CD36/SCARB3 specific IgG was purified by Mouse CD36/SCARB3 affinity chromatography.
Catalog No. 50-241-2494