Learn More
ACROBiosystems Recombinant Protein;Mouse CD38 Protein;HEK293;100UG;CD8,CD38,T10,cADPr hydrolase 1
Supplier:  ACROBiosystems CD8M5223100UG
Encompass_Preferred
Recombinant Protein;Mouse CD38 Protein;100UG;Mouse CD38 (CD8-M5223) is expressed from human 293 cells (HEK293). It contains AA Leu 45 - Thr 304 (Accession # P56528-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 31.7 kDa. The protein migrates as 38-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. NC1536980