Learn More
Sino Biological Mouse CD38 Protein 1MG (Sino Biological)
Supplier:  Sino Biological 50191MNAH1
Encompass_Preferred
A DNA sequence encoding the Mouse CD38 (NP_031672.2) (Leu45-Thr304) was expressed.
Catalog No. 50-237-0651