Learn More
ACROBiosystems ACROBiosystems Mouse CD39 / ENTPD1 Protein, His Tag
Supplier:  ACROBiosystems CD9M52H320UG
Encompass_Preferred
Recombinant Protein;20UG;Mouse CD39, His Tag (CD9-M52H3) is expressed from human 293 cells (HEK293). It contains AA Thr 38 -Ile 478 (Accession # P55772-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 51.7 kDa.
Catalog No. 50-210-6413