Learn More
Sino Biological Recombinant Anti-CD4 Antibody (PE), Rabbit Monoclonal
Supplier:  Sino Biological 50134R711P100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD4 (rM CD4; Catalog#50134-M08H; NP_038516.1; Met1-Thr394) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-241-1085