Learn More
Sino Biological Anti-CD4 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50134T52100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CD4 (rM CD4; Catalog#50134-M08H; NP_038516.1; Met1-Thr394). CD4 specific IgG was purified by Mouse CD4 affinity chromatography.
Catalog No. 50-241-2588