Learn More
ACROBiosystems Recombinant Protein;Mouse CD40 Ligand / TNFSF5 Protein;HEK293;100UG;CDL,CD40LG,CD154,CD40L,HIGM1,IGM,IMD3,T-BAM,TNFSF5,TRAP,gp39,CD40 Ligand,CD40-LG,CD40 L
Supplier:  ACROBiosystems CDLM5248100UG
Encompass_Preferred
Recombinant Protein;Mouse CD40 Ligand / TNFSF5 Protein;100UG;Mouse CD40 Ligand, His Tag (CDL-M5248) is expressed from human 293 cells (HEK293). It contains AA Gly 115 - Leu 260 (Accession # P27548).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 16.8 kDa. The protein migrates as 21-25 kDa under reducing (R) condition (SDS-PAGE).;Immobilized Mouse CD40, Fc Tag (Cat. No. TN5-M5259) at 2 μg/mL ( 100 μl/well ) can bind Mouse CD40 Ligand, His Tag (Cat. No. CDL-M5248) with a linear range of 5-20 ng/mL (QC tested).
Catalog No. 50-113-8293