Learn More
Sino Biological Recombinant Mouse CD40/TNFRSF5 Protein (His & AVI Tag), Biotinylated
Supplier:  Sino Biological 50324M49HB100
Encompass_Preferred
A DNA sequence encoding the mouse CD40 (NP_035741.2) (Met1-Arg193) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-255-3307