Learn More
ACROBiosystems ACROBiosystems Mouse CD40 / TNFRSF5 Protein, Fc Tag
Catalog No. 502106687
Encompass_Preferred
Recombinant Protein;1MG;Mouse CD40, Fc Tag (TN5-M5259) is expressed from human 293 cells (HEK293). It contains AA Val 24 - Arg 193 (Accession # P27512-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 45.0 kDa. The protein migrates as 50-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse CD40, Fc Tag (Cat. No. TN5-M5259) at 2 μg/mL (100 μL/well) can bind Mouse CD40 Ligand, His Tag (Cat. No. CDL-M5248) with a linear range of 9-78 ng/mL (QC tested).
Catalog No. 50-210-6687 Supplier ACROBiosystems Supplier No. TN5M52591MG
Recombinant Protein;1MG;Mouse CD40, Fc Tag (TN5-M5259) is expressed from human 293 cells (HEK293). It contains AA Val 24 - Arg 193 (Accession # P27512-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 45.0 kDa. The protein migrates as 50-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse CD40, Fc Tag (Cat. No. TN5-M5259) at 2 μg/mL (100 μL/well) can bind Mouse CD40 Ligand, His Tag (Cat. No. CDL-M5248) with a linear range of 9-78 ng/mL (QC tested).