Learn More
Biolegend PE/Cyanine7 anti-mouse CD43
Supplier:  Biolegend 143210
Encompass_Preferred
PE/Cyanine7 anti-mouse CD43 [S11]; Isotype: Rat IgG2b; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. NC1314381