Learn More
Raybiotech Inc Mouse CD45 ELISA Kit
Supplier:  Raybiotech Inc ELMCD451
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.26 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Mouse CD45.
Catalog No. 50-194-9946