Learn More
ACROBiosystems Mouse CD47 25ug
Catalog No. 502535338
Encompass_Preferred
Biotinylated Mouse CD47 Protein FcAvitag (CD7-M82F5) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Lys 140 (Accession # Q61735-1).
Catalog No. 50-253-5338 Supplier ACROBiosystems Supplier No. CD7M82F525UG
Biotinylated Mouse CD47 Protein FcAvitag (CD7-M82F5) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Lys 140 (Accession # Q61735-1).