Learn More
ACROBiosystems Recombinant Protein;Mouse CD47 Protein, Fc Tag;HEK293;100UG;CD7,CD47,MER6,IAP,OA3
Supplier:  ACROBiosystems CD7M5251100UG
Encompass_Preferred
Recombinant Protein;Mouse CD47 Protein, Fc Tag;100UG;Mouse CD47, Fc Tag (CD7-M5251) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Lys 140 (Accession # Q61735).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 40 kDa. The protein migrates as 50-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-108-9533