Learn More
Sino Biological Recombinant Mouse CD47 Protein (His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 57231M49HB100
Encompass_Preferred
A DNA sequence encoding the Mouse CD47 (Q61735-1)(Met1-Lys140) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-241-1798