Learn More
Sino Biological Recombinant Mouse CD47 Protein (ECD)
Supplier:  Sino Biological 57231MNAH200
Encompass_Preferred
A DNA sequence encoding the mouse CD47 (ADQ12919.1) (Met1-Lys140) was expressed.
Catalog No. 50-255-2955