Learn More
Sino Biological Mouse CD48 Matched ELISA Antibody Pair Set 5 Plates
Supplier: Sino Biological SEK504155
The Mouse CD48/Blast-1 ELISA Pair Set is for the quantitative determination of Mouse CD48/Blast-1.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Mouse CD48/Blast-1 coated on a 96-well plate. Standards and samples are added to the wells, and any Mouse CD48/Blast-1 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Mouse CD48/Blast-1 monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Mouse CD48/Blast-1 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.