Learn More
ACROBiosystems Biotinylated Mouse CD5 Protein, His,Avitag™
Catalog No. 502331114
Encompass_Preferred
Biotinylated Mouse CD5 Protein, His,Avitag (CD5-M82E3) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Asn 370 (Accession # P13379).
Catalog No. 50-233-1114 Supplier ACROBiosystems Supplier No. CD5M82E325UG
Biotinylated Mouse CD5 Protein, His,Avitag (CD5-M82E3) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Asn 370 (Accession # P13379).