Learn More
Sino Biological Recombinant Anti-CD5 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50403R249100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD5 / Cluster of Differentiation 5 (rM CD5 / Cluster of Differentiation 5; Catalog#50403-M08H; NP_031676.3; Met1-Pro371).
Catalog No. 50-241-2206